missing translation for 'onlineSavingsMsg'
Learn More

1,2-Epoxy-7-octene, 97%

Catalog No. AAL0703306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0703306 5 g
AAL0703314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0703306 Supplier Thermo Scientific Chemicals Supplier No. L0703306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 19600-63-6 | C8H14O | 126.199 g/mol

1,2-Epoxy-7-octene is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Epoxy-7-octene is used as an organic chemical synthesis intermediate.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 19600-63-6
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.199
MDL Number MFCD00005156
InChI Key UKTHULMXFLCNAV-UHFFFAOYSA-N
Synonym 1,2-epoxy-7-octene, oxirane, 5-hexenyl, 5-hexenyloxirane, 7,8-epoxyoctene, ccris 3749, 2-hex-5-en-1-yl oxirane, oxirane, 5-hexenyl-, s, 2-hex-5-en-yloxirane, 2-5-hexenyl oxirane, acmc-20ap64
PubChem CID 29678
IUPAC Name 2-hex-5-enyloxirane
SMILES C=CCCCCC1CO1

Specifications

Density 0.85
Boiling Point 167°C
Flash Point 38°C (100°F)
Refractive Index 1.435
Quantity 5 g
UN Number UN1993
Beilstein 1305290
Solubility Information Not miscible in water.
Formula Weight 126.2
Percent Purity 97%
Chemical Name or Material 1,2-Epoxy-7-octene
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P374-P380-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 243-178-7
RTECSNumber RR0502000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only