missing translation for 'onlineSavingsMsg'
Learn More

1,2-Epoxy-5-hexene 96.0+%, TCI America™

Catalog No. E041025G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E041025G 25 g
E04105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E041025G Supplier TCI America Supplier No. E041025G
Only null left

Chemical Identifiers

CAS 10353-53-4
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
MDL Number MFCD00010051
InChI Key MUUOUUYKIVSIAR-UHFFFAOYSA-N
Synonym 1,2-epoxy-5-hexene, diallyl monoxide, 2-3-butenyl oxirane, diallyl monooxide, oxirane, 3-butenyl, 3-butenyloxirane, 1-hexene, 5,6-epoxy, 2-but-3-en-1-yl oxirane, 5,6-epoxyhex-1-ene, ccris 3748
PubChem CID 66314
IUPAC Name 2-but-3-enyloxirane
SMILES C=CCCC1CO1

Specifications

Color Colorless
Boiling Point 121°C
Quantity 25 g
UN Number 1993
Formula Weight 98.15
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Epoxy-5-hexene
EINECSNumber (5)-6273
RTECSNumber MP6780000
TSCA No