accessibility menu, dialog, popup

UserName

CAS RN 10353-53-4

IUPAC Name: 2-(but-3-en-1-yl)oxirane
Synonyms: 2-(but-3-en-1-yl)oxirane
Molecular Weight (g/mol): 98.15
Molecular Formula: C6H10O
InChi Key: MUUOUUYKIVSIAR-UHFFFAOYNA-N
SMILES: C=CCCC1CO1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)