missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Dodecanediol 93.0+%, TCI America™
$106.25 - $652.41
Chemical Identifiers
| CAS | 1119-87-5 |
|---|---|
| Molecular Formula | C12H26O2 |
| Molecular Weight (g/mol) | 202.34 |
| MDL Number | MFCD00004726 |
| InChI Key | ZITKDVFRMRXIJQ-UHFFFAOYNA-N |
| Synonym | 1,2-Dihydroxydodecane |
| PubChem CID | 92866 |
| IUPAC Name | dodecane-1,2-diol |
| SMILES | CCCCCCCCCCC(O)CO |
Chemical Identifiers
| 1119-87-5 | |
| 202.34 | |
| ZITKDVFRMRXIJQ-UHFFFAOYNA-N | |
| 92866 | |
| CCCCCCCCCCC(O)CO |
| C12H26O2 | |
| MFCD00004726 | |
| 1,2-Dihydroxydodecane | |
| dodecane-1,2-diol |
Specifications
| 1119-87-5 | |
| White | |
| MFCD00004726 | |
| 1,2-Dihydroxydodecane | |
| CCCCCCCCCCC(O)CO | |
| 202.34 | |
| 202.34 | |
| Crystalline Powder |
| 58°C | |
| C12H26O2 | |
| 25 g | |
| ZITKDVFRMRXIJQ-UHFFFAOYNA-N | |
| dodecane-1,2-diol | |
| 92866 | |
| ≥93.0% (GC) | |
| 1,2-Dodecanediol |
Safety and Handling
EINECSNumber : (2)-0240
TSCA : Yes