accessibility menu, dialog, popup

UserName

H3COHOH

CAS RN 1119-87-5

IUPAC Name: dodecane-1,2-diol
Synonyms: lauryl glycol
Molecular Weight (g/mol): 202.34
Molecular Formula: C12H26O2
InChi Key: ZITKDVFRMRXIJQ-UHFFFAOYNA-N
SMILES: CCCCCCCCCCC(O)CO
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results