missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dodecanediol 93.0+%, TCI America™

Catalog No. D186125G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D186125G 25 g
D1861500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D186125G Supplier TCI America Supplier No. D186125G

Chemical Identifiers

CAS 1119-87-5
Molecular Formula C12H26O2
Molecular Weight (g/mol) 202.34
MDL Number MFCD00004726
InChI Key ZITKDVFRMRXIJQ-UHFFFAOYNA-N
Synonym 1,2-Dihydroxydodecane
PubChem CID 92866
IUPAC Name dodecane-1,2-diol
SMILES CCCCCCCCCCC(O)CO

Specifications

Melting Point 58°C
Color White
Quantity 25 g
Formula Weight 202.34
Percent Purity ≥93.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Dodecanediol
EINECSNumber (2)-0240
TSCA Yes