missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dipalmitoyl-sn-glycero-3-phosphocholine 98.0+%, TCI America™

Catalog No. D39251G
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39251G 1 g
D3925200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D39251G Supplier TCI America Supplier No. D39251G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 63-89-8
Molecular Formula C40H80NO8P
Molecular Weight (g/mol) 734.053
MDL Number MFCD00036903
InChI Key KILNVBDSWZSGLL-KXQOOQHDSA-N
Synonym colfosceril palmitate, dppc, 1,2-dipalmitoyl-sn-glycero-3-phosphocholine, 1,2-dipalmitoyl-l-lecithin, dipalmitoylphosphatidylcholine, 1,2-dihexadecanoyl-sn-glycero-3-phosphocholine, colfoscerili palmitas, palmitate de colfosceril, palmitato de colfoscerilo, unii-319x2nfw0a
PubChem CID 452110
ChEBI CHEBI:72999
IUPAC Name [(2R)-2,3-di(hexadecanoyloxy)propyl] 2-(trimethylazaniumyl)ethyl phosphate
SMILES CCCCCCCCCCCCCCCC(=O)OCC(COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CCCCCCCCCCCCCCC

Specifications

Melting Point 220°C
Color White
Quantity 1 g
Formula Weight 734.05
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Dipalmitoyl-sn-glycero-3-phosphocholine
TSCA No
Recommended Storage Freezer