accessibility menu, dialog, popup

UserName

CAS RN 63-89-8

IUPAC Name: (2-{[2,3-bis(hexadecanoyloxy)propyl phosphonato]oxy}ethyl)trimethylazanium
Synonyms: colfosceril palmitate dipalmitoyl glycerophosphocholine
Molecular Weight (g/mol): 734.05
Molecular Formula: C40H80NO8P
InChi Key: KILNVBDSWZSGLL-UHFFFAOYNA-N
SMILES: CCCCCCCCCCCCCCCC(=O)OCC(COP([O-])(=O)OCC[N+](C)(C)C)OC(=O)CCCCCCCCCCCCCCC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results