missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dimethylnaphthalene 95.0+%, TCI America™

Catalog No. D07471G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D07471G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D07471G Supplier TCI America Supplier No. D07471G

Chemical Identifiers

CAS 573-98-8
Molecular Formula C12H12
Molecular Weight (g/mol) 156.228
MDL Number MFCD00004035
InChI Key QNLZIZAQLLYXTC-UHFFFAOYSA-N
Synonym dimethylnaphthalene, naphthalene, dimethyl, naphthalene, 1,2-dimethyl, 1,2-dimethyl naphthalene, unii-23t7o135bd, 1,2-dmn, 1,2-dimethyl-naphthalene, dsstox_cid_5130, dsstox_rid_77675, dsstox_gsid_25130
PubChem CID 11317
ChEBI CHEBI:34052
IUPAC Name 1,2-dimethylnaphthalene
SMILES CC1=C(C2=CC=CC=C2C=C1)C

Specifications

Melting Point -2°C
Color Yellow
Boiling Point 132°C
Quantity 1 g
Formula Weight 156.23
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Dimethylnaphthalene