Learn More
1,2-Dimethylimidazole, 98%
CAS: 1739-84-0 | C5H8N2 | 96.13 g/mol
$42.01 - $231.26
Chemical Identifiers
| CAS | 1739-84-0 |
|---|---|
| Molecular Formula | C5H8N2 |
| Molecular Weight (g/mol) | 96.13 |
| MDL Number | MFCD00005294 |
| InChI Key | GIWQSPITLQVMSG-UHFFFAOYSA-N |
| Synonym | 1,2-dimethyl-1h-imidazole, 1h-imidazole, 1,2-dimethyl, imidazole, 1,2-dimethyl, 1h-imidazole, dimethyl, unii-q41bc3grjb, q41bc3grjb, lupragen r dmi, 1,2-dimethylimdazole, 1,2 dimethylimidazole, n,2-dimethylimidazole |
| PubChem CID | 15617 |
| IUPAC Name | 1,2-dimethylimidazole |
| SMILES | CN1C=CN=C1C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2354414
|
Thermo Scientific Chemicals
B2354414 |
25 g |
Each for $42.01
|
|
|||||
|
AAB2354422
|
Thermo Scientific Chemicals
B2354422 |
100 g |
Each for $88.10
|
|
|||||
|
AAB2354436
|
Thermo Scientific Chemicals
B2354436 |
500 g |
Each for $231.26
|
|
|||||
Description
1,2-Dimethylimidazole is used in the synthesis of 1,2-dimethyl-3-n-butylimidazoliumchloride and 1,2-dimethyl-3-n-propylimidazolium chloride. It also can be used in the synthesis of 1-(2-methoxyethyl)-2,3-dimethylimidazolium chloride and hexafluorophosphate salts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,2-Dimethylimidazole is used in the synthesis of 1,2-dimethyl-3-n-butylimidazoliumchloride and 1,2-dimethyl-3-n-propylimidazolium chloride. It also can be used in the synthesis of 1-(2-methoxyethyl)-2,3-dimethylimidazolium chloride and hexafluorophosphate salts.
Solubility
Soluble in water.
Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Chemical Identifiers
| 1739-84-0 | |
| 96.13 | |
| GIWQSPITLQVMSG-UHFFFAOYSA-N | |
| 15617 | |
| CN1C=CN=C1C |
| C5H8N2 | |
| MFCD00005294 | |
| 1,2-dimethyl-1h-imidazole, 1h-imidazole, 1,2-dimethyl, imidazole, 1,2-dimethyl, 1h-imidazole, dimethyl, unii-q41bc3grjb, q41bc3grjb, lupragen r dmi, 1,2-dimethylimdazole, 1,2 dimethylimidazole, n,2-dimethylimidazole | |
| 1,2-dimethylimidazole |
Specifications
| 1739-84-0 | |
| 1.084 | |
| 92°C (197°F) | |
| MFCD00005294 | |
| 108457 | |
| Soluble in water. | |
| CN1C=CN=C1C | |
| 96.13 | |
| 96.13 | |
| 1,2-Dimethylimidazole |
| 35°C to 38°C | |
| 203°C to 205°C | |
| C5H8N2 | |
| 25 g | |
| 1,2-dimethyl-1h-imidazole, 1h-imidazole, 1,2-dimethyl, imidazole, 1,2-dimethyl, 1h-imidazole, dimethyl, unii-q41bc3grjb, q41bc3grjb, lupragen r dmi, 1,2-dimethylimdazole, 1,2 dimethylimidazole, n,2-dimethylimidazole | |
| GIWQSPITLQVMSG-UHFFFAOYSA-N | |
| 1,2-dimethylimidazole | |
| 15617 | |
| 98% |
Safety and Handling
GHS H Statement
H318-H302-H315
Causes serious eye damage.
Harmful if swallowed.
Causes skin irritation.
P210-P234-P235-P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P333+P313-P362-P370+P378q-P390-P501c
H227-H302-H315-H317-H318-H335
EINECSNumber : 217-101-2
RTECSNumber : NI4838854
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only