Learn More
1,2-Dimethylimidazole, 98%
CAS: 1739-84-0 | C5H8N2 | 96.13 g/mol
Supplier: Thermo Scientific Chemicals B2354422
Description
1,2-Dimethylimidazole is used in the synthesis of 1,2-dimethyl-3-n-butylimidazoliumchloride and 1,2-dimethyl-3-n-propylimidazolium chloride. It also can be used in the synthesis of 1-(2-methoxyethyl)-2,3-dimethylimidazolium chloride and hexafluorophosphate salts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,2-Dimethylimidazole is used in the synthesis of 1,2-dimethyl-3-n-butylimidazoliumchloride and 1,2-dimethyl-3-n-propylimidazolium chloride. It also can be used in the synthesis of 1-(2-methoxyethyl)-2,3-dimethylimidazolium chloride and hexafluorophosphate salts.
Solubility
Soluble in water.
Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications
| 1,2-Dimethylimidazole | |
| 35°C to 38°C | |
| 203°C to 205°C | |
| C5H8N2 | |
| 100 g | |
| 1,2-dimethyl-1h-imidazole, 1h-imidazole, 1,2-dimethyl, imidazole, 1,2-dimethyl, 1h-imidazole, dimethyl, unii-q41bc3grjb, q41bc3grjb, lupragen r dmi, 1,2-dimethylimdazole, 1,2 dimethylimidazole, n,2-dimethylimidazole | |
| GIWQSPITLQVMSG-UHFFFAOYSA-N | |
| 1,2-dimethylimidazole | |
| 15617 | |
| 98% |
| 1739-84-0 | |
| 1.084 | |
| 92°C (197°F) | |
| MFCD00005294 | |
| 108457 | |
| Soluble in water. | |
| CN1C=CN=C1C | |
| 96.13 | |
| 96.13 |
Chemical Identifiers
| 1739-84-0 | |
| 96.13 | |
| GIWQSPITLQVMSG-UHFFFAOYSA-N | |
| 15617 | |
| CN1C=CN=C1C |
| C5H8N2 | |
| MFCD00005294 | |
| 1,2-dimethyl-1h-imidazole, 1h-imidazole, 1,2-dimethyl, imidazole, 1,2-dimethyl, 1h-imidazole, dimethyl, unii-q41bc3grjb, q41bc3grjb, lupragen r dmi, 1,2-dimethylimdazole, 1,2 dimethylimidazole, n,2-dimethylimidazole | |
| 1,2-dimethylimidazole |
Safety and Handling
GHS H Statement
H318-H302-H315
Causes serious eye damage.
Harmful if swallowed.
Causes skin irritation.
P210-P234-P235-P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P333+P313-P362-P370+P378q-P390-P501c
H227-H302-H315-H317-H318-H335
EINECSNumber : 217-101-2
RTECSNumber : NI4838854
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only