missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Dimethoxyethane 99.0+%, TCI America™
Supplier: TCI America D0634500ML
Specifications
| 1,2-Dimethoxyethane | |
| Colorless | |
| C4H10O2 | |
| 500 mL | |
| monoglyme, ethylene glycol dimethyl ether, glyme, dimethyl cellosolve, egdme, ethane, 1,2-dimethoxy, dimethoxyethane, 2,5-dioxahexane, glycol dimethyl ether, ethylene dimethyl ether | |
| COCCOC | |
| 90.122 | |
| CHEBI:42263 | |
| ≥99.0% (GC) |
| 110-71-4 | |
| 84°C | |
| MFCD00008502 | |
| 2252 | |
| XTHFKEDIFFGKHM-UHFFFAOYSA-N | |
| 1,2-dimethoxyethane | |
| 8071 | |
| 90.12 | |
| Liquid |
Chemical Identifiers
| 110-71-4 | |
| 90.122 | |
| XTHFKEDIFFGKHM-UHFFFAOYSA-N | |
| 8071 | |
| 1,2-dimethoxyethane |
| C4H10O2 | |
| MFCD00008502 | |
| monoglyme, ethylene glycol dimethyl ether, glyme, dimethyl cellosolve, egdme, ethane, 1,2-dimethoxy, dimethoxyethane, 2,5-dioxahexane, glycol dimethyl ether, ethylene dimethyl ether | |
| CHEBI:42263 | |
| COCCOC |
Safety and Handling
EINECSNumber : (2)-0421&(7)-1321
RTECSNumber : KI1451000
TSCA : Yes