missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dimethoxyethane 99.0+%, TCI America™

Catalog No. D063425ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D063425ML 25 mL
D0634500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D063425ML Supplier TCI America Supplier No. D063425ML

Chemical Identifiers

CAS 110-71-4
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.122
MDL Number MFCD00008502
InChI Key XTHFKEDIFFGKHM-UHFFFAOYSA-N
Synonym monoglyme, ethylene glycol dimethyl ether, glyme, dimethyl cellosolve, egdme, ethane, 1,2-dimethoxy, dimethoxyethane, 2,5-dioxahexane, glycol dimethyl ether, ethylene dimethyl ether
PubChem CID 8071
ChEBI CHEBI:42263
IUPAC Name 1,2-dimethoxyethane
SMILES COCCOC

Specifications

Color Colorless
Boiling Point 84°C
Quantity 25 mL
UN Number 2252
Formula Weight 90.12
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Dimethoxyethane
EINECSNumber (2)-0421&(7)-1321
RTECSNumber KI1451000
TSCA Yes