missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dimethoxyethane, Reagent, 99.5%, Spectrum™ Chemical

Catalog No. p-7157357
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600031 100 mL Amber Glass Bottle
18600032 500 mL Amber Glass Bottle
18600033 1 L Amber Glass Bottle
18600034 4 L Amber Glass Bottle
18600035 4 x 4 L Amber Glass Bottle
18600036 20 L Steel Can
18600037 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18600031 Supplier Spectrum Chemical Mfg Cor Supplier No. DI139100ML
Only null left

DI139, 110-71-4, C4H10O2

Spectrum™ Chemical 1,2-Dimethoxyethane, Reagent is also known as glyme, monoglyme, dimethyl glycol, ethylene glycol dimethyl ether, dimethyl cellosolve and DME. Dimethoxyethane is a clear, colorless, aprotic liquid ether. It is commonly used as a solvent in batteries and in organometallic reaction chemistry since it is higher boiling than diethyl ether and THF. It is also miscible with water. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 110-71-4
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
InChI Key XTHFKEDIFFGKHM-UHFFFAOYSA-N
IUPAC Name 1,2-dimethoxyethane
SMILES COCCOC

Specifications

Acidity 0.015%
CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 100 mL
UN Number UN2252
Percent Purity 99.5%
Grade Reagent