missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dimethoxyethane, 99+%, stab. with BHT

Catalog No. p-7020879
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA12986AE 100 mL
AAA12986AP 500 mL
AAA129860F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAA12986AE Supplier Thermo Scientific Chemicals Supplier No. A12986AE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 110-71-4 | C4H10O2 | 90.122 g/mol

1,2-Dimethoxyethane is widely used as a solvent for electrolyte of lithium batteries, polysilicones, oligo- and polysaccharides. It plays an important role in Grignard reactions, Suzuki reactions and Stille couplings in organometallic chemistry and in pharmaceutical synthesis. It is a higher boiling point solvent and is used as an alternative to diethyl ether and tetrahydrofuran. It is used for the etching of synthetic polymers like polytetrafluoroethylene and other fluoropolymers with alkali metal dispersions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Dimethoxyethane is widely used as a solvent for electrolyte of lithium batteries, polysilicones, oligo- and polysaccharides. It plays an important role in Grignard reactions, Suzuki reactions and Stille couplings in organometallic chemistry and in pharmaceutical synthesis. It is a higher boiling point solvent and is used as an alternative to diethyl ether and tetrahydrofuran. It is used for the etching of synthetic polymers like polytetrafluoroethylene and other fluoropolymers with alkali metal dispersions.

Solubility
Miscible with water, methanol, ethanol, diethyl ether, acetone, tetrahydrofuran, benzene and toluene.

Notes
Keep away from sources of ignition. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents. It can be oxidized on air to form peroxides.

Chemical Identifiers

CAS 110-71-4
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.122
MDL Number MFCD00008502
InChI Key XTHFKEDIFFGKHM-UHFFFAOYSA-N
Synonym monoglyme, ethylene glycol dimethyl ether, glyme, dimethyl cellosolve, egdme, ethane, 1,2-dimethoxy, dimethoxyethane, 2,5-dioxahexane, glycol dimethyl ether, ethylene dimethyl ether
PubChem CID 8071
ChEBI CHEBI:42263
IUPAC Name 1,2-dimethoxyethane
SMILES COCCOC

Specifications

Melting Point -69°C
Density 0.867
Boiling Point 84°C to 86°C
Flash Point −6°C (21°F)
Odor Ether-like
Linear Formula CH3OCH2CH2OCH3
Refractive Index 1.38
Quantity 100 mL
UN Number UN2252
Beilstein 1209237
Merck Index 14,3224
Solubility Information Miscible with water,methanol,ethanol,diethyl ether,acetone,tetrahydrofuran,benzene and toluene.
Formula Weight 90.12
Percent Purity ≥99%
Chemical Name or Material 1,2-Dimethoxyethane, stabilized with BHT
Show More Show Less
Hazard Category H225-H315-H319-H332-H360FD
Hazard Statement GHS H Statement
H225-H360-H332
Highly flammable liquid and vapour.
May damage fertility or the unborn child.
Harmful if inhaled.
Precautionary Statement P201-P202-P210-P233-P240-P241-P242-P243-P261-P264b-P271-P280i-P281-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 1,2-DIMETHOXYETHANE
EINECSNumber 203-794-9
RTECSNumber KI1451000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only