missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dimethoxyethane, ≥99%, Reagent Grade, Inhibitor-free, Honeywell™

Catalog No. p-7120016
Change view
Click to view available options
Quantity:
1 L
2.5 L
20 L
Packaging:
Glass bottle
Stainless steel can
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60047983 1 L Glass bottle
60047984 2.5 L Glass bottle
60047985 20 L Stainless steel can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60047983 Supplier Honeywell Chemicals Supplier No. E274081L
Only null left

Reagent Grade, inhibitor-free,≥99%

  • ≥95% for general laboratory use.

Chemical Identifiers

CAS 110-71-4
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.122
MDL Number MFCD00008502
InChI Key XTHFKEDIFFGKHM-UHFFFAOYSA-N
Synonym monoglyme, ethylene glycol dimethyl ether, glyme, dimethyl cellosolve, egdme, ethane, 1,2-dimethoxy, dimethoxyethane, 2,5-dioxahexane, glycol dimethyl ether, ethylene dimethyl ether
PubChem CID 8071
ChEBI CHEBI:42263
IUPAC Name 1,2-dimethoxyethane
SMILES COCCOC

Specifications

Melting Point -58°C
Color Colorless
Density 0.867g/mL at 25°C
pH Neutral
Boiling Point 85°C (1013 hPa)
Flash Point −2°C (28.4°F)
Packaging Glass bottle
Linear Formula CH3OCH2CH2OCH3
Quantity 1 L
UN Number UN2252
Beilstein 1209237
Vapor Density 3.1 (20°C, vs air)
Formula Weight 90.12g/mol
Percent Purity ≥99%
Chemical Name or Material 1,2-Dimethoxyethane
Show More Show Less
Hazard Statement P201-P210-P280-P308 + P313-P370 + P378-P403 + P235
DOTInformation 3/PGII
RTECSNumber 203-794-9