missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Dimethoxybenzene 99.0+%, TCI America™
Supplier: TCI America D0627500G
Specifications
| 1,2-Dimethoxybenzene | |
| White | |
| C8H10O2 | |
| 500 g | |
| ABDKAPXRBAPSQN-UHFFFAOYSA-N | |
| 1,2-dimethoxybenzene | |
| 7043 | |
| 138.17 | |
| Crystalline Lumps |
| 91-16-7 | |
| 207°C | |
| MFCD00008357 | |
| veratrole, veratrol, pyrocatechol dimethyl ether, o-dimethoxybenzene, catechol dimethyl ether, benzene, 1,2-dimethoxy, 2-methoxyanisole, o,o-dimethyl catechol, 2-dimethoxybenzol, benzene, o-dimethoxy | |
| COC1=CC=CC=C1OC | |
| 138.166 | |
| CHEBI:59114 | |
| ≥99.0% (GC) |
Chemical Identifiers
| 91-16-7 | |
| 138.166 | |
| ABDKAPXRBAPSQN-UHFFFAOYSA-N | |
| 7043 | |
| 1,2-dimethoxybenzene |
| C8H10O2 | |
| MFCD00008357 | |
| veratrole, veratrol, pyrocatechol dimethyl ether, o-dimethoxybenzene, catechol dimethyl ether, benzene, 1,2-dimethoxy, 2-methoxyanisole, o,o-dimethyl catechol, 2-dimethoxybenzol, benzene, o-dimethoxy | |
| CHEBI:59114 | |
| COC1=CC=CC=C1OC |
Safety and Handling
EINECSNumber : (3)-0582
RTECSNumber : CZ6475000
TSCA : Yes