missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dimethoxybenzene 99.0+%, TCI America™

Catalog No. D062725G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D062725G 25 g
D0627100G 100 g
D0627500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D062725G Supplier TCI America Supplier No. D062725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 91-16-7
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00008357
InChI Key ABDKAPXRBAPSQN-UHFFFAOYSA-N
Synonym veratrole, veratrol, pyrocatechol dimethyl ether, o-dimethoxybenzene, catechol dimethyl ether, benzene, 1,2-dimethoxy, 2-methoxyanisole, o,o-dimethyl catechol, 2-dimethoxybenzol, benzene, o-dimethoxy
PubChem CID 7043
ChEBI CHEBI:59114
IUPAC Name 1,2-dimethoxybenzene
SMILES COC1=CC=CC=C1OC

Specifications

Color White
Boiling Point 207°C
Quantity 25 g
Formula Weight 138.17
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1,2-Dimethoxybenzene
EINECSNumber (3)-0582
RTECSNumber CZ6475000
TSCA Yes