missing translation for 'onlineSavingsMsg'
Learn More

1,2-Diiodoethane, 98%

CAS: 624-73-7 | C2H4I2 | 281.863 g/mol

Supplier:  Thermo Scientific Chemicals L0301906

 View more versions of this product

Catalog No. AAL0301906


Only null left
Explore available promotions

Description

Description

1,2-Diiodoethane is used as C-X bond formation reagent. It is also used as Pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Diiodoethane is used as C-X bond formation reagent. It is also used as Pharmaceutical intermediates.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Store at 4°C. Air & Light Sensitive. Incompatible with strong bases, strong oxidizing agents, heat and light. Protect from heat.
Specifications

Specifications

1,2-Diiodoethane
80°C to 84°C
C2H4I2
5 g
Light sensitive
Soluble in methanol. Insoluble in water.
C(CI)I
281.863
281.86
624-73-7
2.13
MFCD00001093
1730870
ethane, 1,2-diiodo, ethylene diiodide, ethylene iodide, 1,2-diiodethane, 1,2-diiodo ethane, 1,2-bis iodanyl ethane, ch2ich2i, 1,2-diiodoethane, acmc-1b2u1, ksc358c5b
GBBZLMLLFVFKJM-UHFFFAOYSA-N
1,2-diiodoethane
12224
98%

Chemical Identifiers

624-73-7
281.863
GBBZLMLLFVFKJM-UHFFFAOYSA-N
12224
C(CI)I
C2H4I2
MFCD00001093
ethane, 1,2-diiodo, ethylene diiodide, ethylene iodide, 1,2-diiodethane, 1,2-diiodo ethane, 1,2-bis iodanyl ethane, ch2ich2i, 1,2-diiodoethane, acmc-1b2u1, ksc358c5b
1,2-diiodoethane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 210-859-5

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only