missing translation for 'onlineSavingsMsg'
Learn More

1,2-Diiodobenzene (stabilized with Copper chip) 98.0+%, TCI America™

Catalog No. p-7126316
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D06065G 5 g
D060625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D06065G Supplier TCI America Supplier No. D06065G
Only null left

Chemical Identifiers

CAS 615-42-9
Molecular Formula C6H4I2
Molecular Weight (g/mol) 329.91
MDL Number MFCD00001038
InChI Key BBOLNFYSRZVALD-UHFFFAOYSA-N
Synonym o-diiodobenzene, benzene, 1,2-diiodo, benzene, o-diiodo, diiodobenzene, benzene, diiodo, benzene,2-diiodo, pubchem3831, 1,2-diiodobenzene
PubChem CID 11994
IUPAC Name 1,2-diiodobenzene
SMILES IC1=CC=CC=C1I

Specifications

Color Yellow
Boiling Point 152°C
Quantity 5 g
Formula Weight 329.91
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1,2-Diiodobenzene (stabilized with Copper chip)
TSCA Yes
Recommended Storage Refrigerator