missing translation for 'onlineSavingsMsg'
Learn More

1,2-Diiodobenzene, 98%

Catalog No. AC218400050
Change view
Click to view available options
Quantity:
5 g
20 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC218400050 5 g Glass bottle
AC218400200 20 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC218400050 Supplier Thermo Scientific Chemicals Supplier No. 218400050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 615-42-9 | C6H4I2 | 329.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 615-42-9
Molecular Formula C6H4I2
Molecular Weight (g/mol) 329.91
MDL Number MFCD00001038
InChI Key BBOLNFYSRZVALD-UHFFFAOYSA-N
Synonym o-diiodobenzene, benzene, 1,2-diiodo, benzene, o-diiodo, diiodobenzene, benzene, diiodo, benzene,2-diiodo, pubchem3831, 1,2-diiodobenzene
PubChem CID 11994
IUPAC Name 1,2-diiodobenzene
SMILES IC1=CC=CC=C1I

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Brown to Yellow
Density 2.5200g/mL
Boiling Point 152°C (15.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.7176
Linear Formula C6H4I2
Quantity 5 g
Beilstein 05, 225
Specific Gravity 2.52
Formula Weight 329.9
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1, 2-Diiodobenzene
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye damage.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 210-425-5

RUO – Research Use Only