missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dihydroxynaphthalene 95.0+%, TCI America™

Catalog No. p-7127216
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22991G 1 g
D22995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D22991G Supplier TCI America Supplier No. D22991G

Chemical Identifiers

CAS 574-00-5
Molecular Formula C10H8O2
Molecular Weight (g/mol) 160.172
MDL Number MFCD00003959
InChI Key NXPPAOGUKPJVDI-UHFFFAOYSA-N
Synonym 1,2-Naphthalenediol
PubChem CID 11318
ChEBI CHEBI:17435
IUPAC Name naphthalene-1,2-diol
SMILES C1=CC=C2C(=C1)C=CC(=C2O)O

Specifications

Melting Point 103°C
Color Red-White
Quantity 1 g
Formula Weight 160.17
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Dihydroxynaphthalene
EINECSNumber (4)-0356
TSCA No