missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Difluorobenzene 98.0+%, TCI America™
$54.06 - $150.98
Chemical Identifiers
| CAS | 367-11-3 |
|---|---|
| Molecular Formula | C6H4F2 |
| Molecular Weight (g/mol) | 114.10 |
| MDL Number | MFCD00000284 |
| InChI Key | GOYDNIKZWGIXJT-UHFFFAOYSA-N |
| Synonym | o-difluorobenzene, benzene, 1,2-difluoro, benzene, o-difluoro, ortho-difluorobenzene, o-difluoro benzene, difluorobenzene, 1,2-difluoro-benzene, unii-aw7qgmw29c, aw7qgmw29c, 0-difluorobenzene |
| PubChem CID | 9706 |
| ChEBI | CHEBI:38583 |
| IUPAC Name | 1,2-difluorobenzene |
| SMILES | FC1=CC=CC=C1F |
Chemical Identifiers
| 367-11-3 | |
| 114.10 | |
| GOYDNIKZWGIXJT-UHFFFAOYSA-N | |
| 9706 | |
| 1,2-difluorobenzene |
| C6H4F2 | |
| MFCD00000284 | |
| o-difluorobenzene, benzene, 1,2-difluoro, benzene, o-difluoro, ortho-difluorobenzene, o-difluoro benzene, difluorobenzene, 1,2-difluoro-benzene, unii-aw7qgmw29c, aw7qgmw29c, 0-difluorobenzene | |
| CHEBI:38583 | |
| FC1=CC=CC=C1F |
Specifications
| 367-11-3 | |
| 91°C | |
| MFCD00000284 | |
| 1993 | |
| GOYDNIKZWGIXJT-UHFFFAOYSA-N | |
| 1,2-difluorobenzene | |
| 9706 | |
| 114.09 | |
| Liquid |
| Yellow | |
| C6H4F2 | |
| 5 g | |
| o-difluorobenzene, benzene, 1,2-difluoro, benzene, o-difluoro, ortho-difluorobenzene, o-difluoro benzene, difluorobenzene, 1,2-difluoro-benzene, unii-aw7qgmw29c, aw7qgmw29c, 0-difluorobenzene | |
| FC1=CC=CC=C1F | |
| 114.10 | |
| CHEBI:38583 | |
| ≥98.0% (GC) | |
| 1,2-Difluorobenzene |
Safety and Handling
EINECSNumber : (3)-2688
RTECSNumber : CZ5655000
TSCA : No