missing translation for 'onlineSavingsMsg'
Learn More

1,2-Difluorobenzene 98.0+%, TCI America™

Catalog No. D16275G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D16275G 5 g
D162725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D16275G Supplier TCI America Supplier No. D16275G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 367-11-3
Molecular Formula C6H4F2
Molecular Weight (g/mol) 114.10
MDL Number MFCD00000284
InChI Key GOYDNIKZWGIXJT-UHFFFAOYSA-N
Synonym o-difluorobenzene, benzene, 1,2-difluoro, benzene, o-difluoro, ortho-difluorobenzene, o-difluoro benzene, difluorobenzene, 1,2-difluoro-benzene, unii-aw7qgmw29c, aw7qgmw29c, 0-difluorobenzene
PubChem CID 9706
ChEBI CHEBI:38583
IUPAC Name 1,2-difluorobenzene
SMILES FC1=CC=CC=C1F

Specifications

Color Yellow
Boiling Point 91°C
Quantity 5 g
UN Number 1993
Formula Weight 114.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Difluorobenzene
EINECSNumber (3)-2688
RTECSNumber CZ5655000
TSCA No