Learn More
1,2-Difluorobenzene, 98+%
CAS: 367-11-3 | C6H4F2 | 114.10 g/mol
$102.74 - $1159.81
Chemical Identifiers
| CAS | 367-11-3 |
|---|---|
| Molecular Formula | C6H4F2 |
| Molecular Weight (g/mol) | 114.10 |
| MDL Number | MFCD00000284 |
| InChI Key | GOYDNIKZWGIXJT-UHFFFAOYSA-N |
| Synonym | o-difluorobenzene, benzene, 1,2-difluoro, benzene, o-difluoro, ortho-difluorobenzene, o-difluoro benzene, difluorobenzene, 1,2-difluoro-benzene, unii-aw7qgmw29c, aw7qgmw29c, 0-difluorobenzene |
| PubChem CID | 9706 |
| ChEBI | CHEBI:38583 |
| IUPAC Name | 1,2-difluorobenzene |
| SMILES | FC1=CC=CC=C1F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1037709
|
Thermo Scientific Chemicals
A1037709 |
10 g |
Each for $102.74
|
|
|||||
|
AAA1037718
|
Thermo Scientific Chemicals
A1037718 |
50 g |
Each for $301.86
|
|
|||||
|
AAA1037730
|
Thermo Scientific Chemicals
A1037730 |
250 g |
Each for $1,159.81
|
|
|||||
Description
1,2-Difluorobenzene(1,2-DFB) has been used to study the mechanism of dissociation of o-, m- and p-difluorobenzene ions by threshold photoelectron photoion coincidence spectroscopy. It has been used to study the room temperature adsorption of 1,2-DFB, 1,2-dichlorobenzene and 1,2-dibromobenzene on Si(100)2x1 by X-ray photoelectron spectroscopy and temperature programmed desorption. It is used as solvent in electrochemical studies on transition metal complexes. It is also applied as a solvent useful for electrochemical studies on transition metal complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,2-Difluorobenzene(1,2-DFB) has been used to study the mechanism of dissociation of o-, m- and p-difluorobenzene ions by threshold photoelectron photoion coincidence spectroscopy. It has been used to study the room temperature adsorption of 1,2-DFB, 1,2-dichlorobenzene and 1,2-dibromobenzene on Si(100)2x1 by X-ray photoelectron spectroscopy and temperature programmed desorption. It is used as solvent in electrochemical studies on transition metal complexes. It is also applied as a solvent useful for electrochemical studies on transition metal complexes.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources. Protect against electrostatic charges.
Chemical Identifiers
| 367-11-3 | |
| 114.10 | |
| GOYDNIKZWGIXJT-UHFFFAOYSA-N | |
| 9706 | |
| 1,2-difluorobenzene |
| C6H4F2 | |
| MFCD00000284 | |
| o-difluorobenzene, benzene, 1,2-difluoro, benzene, o-difluoro, ortho-difluorobenzene, o-difluoro benzene, difluorobenzene, 1,2-difluoro-benzene, unii-aw7qgmw29c, aw7qgmw29c, 0-difluorobenzene | |
| CHEBI:38583 | |
| FC1=CC=CC=C1F |
Specifications
| 367-11-3 | |
| 1.171 | |
| 1°C (33°F) | |
| C6H4F2 | |
| MFCD00000284 | |
| UN1993 | |
| o-difluorobenzene, benzene, 1,2-difluoro, benzene, o-difluoro, ortho-difluorobenzene, o-difluoro benzene, difluorobenzene, 1,2-difluoro-benzene, unii-aw7qgmw29c, aw7qgmw29c, 0-difluorobenzene | |
| GOYDNIKZWGIXJT-UHFFFAOYSA-N | |
| 1,2-difluorobenzene | |
| 9706 | |
| 114.09 | |
| 1,2-Difluorobenzene |
| -34°C | |
| 92°C to 94°C | |
| Characteristic | |
| 1.444 | |
| 10 g | |
| 1905113 | |
| Not miscible or difficult to mix in water. | |
| FC1=CC=CC=C1F | |
| 114.10 | |
| CHEBI:38583 | |
| ≥98% |
Safety and Handling
GHS H Statement
H225-H319
Highly flammable liquid and vapor.
Causes serious eye irritation.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H225
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 206-680-7
RTECSNumber : CZ5655000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only