missing translation for 'onlineSavingsMsg'
Learn More

1,2-Difluorobenzene, 98+%

Catalog No. AAA1037730
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1037730 250 g
AAA1037709 10 g
AAA1037718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1037730 Supplier Thermo Scientific Chemicals Supplier No. A1037730
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 367-11-3 | C6H4F2 | 114.10 g/mol

1,2-Difluorobenzene(1,2-DFB) has been used to study the mechanism of dissociation of o-, m- and p-difluorobenzene ions by threshold photoelectron photoion coincidence spectroscopy. It has been used to study the room temperature adsorption of 1,2-DFB, 1,2-dichlorobenzene and 1,2-dibromobenzene on Si(100)2x1 by X-ray photoelectron spectroscopy and temperature programmed desorption. It is used as solvent in electrochemical studies on transition metal complexes. It is also applied as a solvent useful for electrochemical studies on transition metal complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Difluorobenzene(1,2-DFB) has been used to study the mechanism of dissociation of o-, m- and p-difluorobenzene ions by threshold photoelectron photoion coincidence spectroscopy. It has been used to study the room temperature adsorption of 1,2-DFB, 1,2-dichlorobenzene and 1,2-dibromobenzene on Si(100)2x1 by X-ray photoelectron spectroscopy and temperature programmed desorption. It is used as solvent in electrochemical studies on transition metal complexes. It is also applied as a solvent useful for electrochemical studies on transition metal complexes.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources. Protect against electrostatic charges.

Chemical Identifiers

CAS 367-11-3
Molecular Formula C6H4F2
Molecular Weight (g/mol) 114.10
MDL Number MFCD00000284
InChI Key GOYDNIKZWGIXJT-UHFFFAOYSA-N
Synonym o-difluorobenzene, benzene, 1,2-difluoro, benzene, o-difluoro, ortho-difluorobenzene, o-difluoro benzene, difluorobenzene, 1,2-difluoro-benzene, unii-aw7qgmw29c, aw7qgmw29c, 0-difluorobenzene
PubChem CID 9706
ChEBI CHEBI:38583
IUPAC Name 1,2-difluorobenzene
SMILES FC1=CC=CC=C1F

Specifications

Melting Point -34°C
Density 1.171
Boiling Point 92°C to 94°C
Flash Point 1°C (33°F)
Odor Characteristic
Refractive Index 1.444
Quantity 250 g
UN Number UN1993
Beilstein 1905113
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 114.09
Percent Purity ≥98%
Chemical Name or Material 1,2-Difluorobenzene
Show More Show Less
Hazard Category H225
Hazard Statement GHS H Statement
H225-H319
Highly flammable liquid and vapor.
Causes serious eye irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 206-680-7
RTECSNumber CZ5655000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only