missing translation for 'onlineSavingsMsg'
Learn More

1,2-Difluoro-4-[trans-4-[2-(trans-4-propylcyclohexyl)ethyl]cyclohexyl]benzene 98.0+%, TCI America™

Catalog No. p-7128665
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D51281G 1 g
D51285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D51281G Supplier TCI America Supplier No. D51281G
Only null left

Chemical Identifiers

CAS 117943-37-0
Molecular Formula C23H34F2
Molecular Weight (g/mol) 348.52
MDL Number MFCD12828126
InChI Key HTAPXFSKUGZEEP-UHFFFAOYSA-N
PubChem CID 587775
IUPAC Name 1,2-difluoro-4-{4-[2-(4-propylcyclohexyl)ethyl]cyclohexyl}benzene
SMILES CCCC1CCC(CCC2CCC(CC2)C2=CC(F)=C(F)C=C2)CC1

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 348.52
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Difluoro-4-[trans-4-[2-(trans-4-propylcyclohexyl)ethyl]cyclohexyl]benzene