missing translation for 'onlineSavingsMsg'
Learn More

1,2-Difluoro-4-nitrobenzene, 98+%

Catalog No. AAA1106009
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1106009 10 g
AAA1106018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1106009 Supplier Thermo Scientific Chemicals Supplier No. A1106009
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 369-34-6 | C6H3F2NO2 | 159.092 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 369-34-6
Molecular Formula C6H3F2NO2
Molecular Weight (g/mol) 159.092
MDL Number MFCD00007198
InChI Key RUBQQRMAWLSCCJ-UHFFFAOYSA-N
Synonym 3,4-difluoronitrobenzene, benzene, 1,2-difluoro-4-nitro, 1-nitro-3,4-difluorobenzene, 1,2-difluoro-4-nitro-benzene, 3,4-difluoro nitrobenzene, pubchem2290, 3,4-difluornitrobenzene, 3.4-difluoronitrobenzene, 3, 4-difluoronitrobenzene, 3,4-difluoro-nitrobenzene
PubChem CID 123053
IUPAC Name 1,2-difluoro-4-nitrobenzene
SMILES C1=CC(=C(C=C1[N+](=O)[O-])F)F

Specifications

Density 1.441
Boiling Point 76°C to 80°C (8 mmHg)
Flash Point 80°C (176°F)
Refractive Index 1.508
Quantity 10 g
UN Number UN2810
Beilstein 1944996
Formula Weight 159.09
Percent Purity ≥98%
Chemical Name or Material 1,2-Difluoro-4-nitrobenzene
Hazard Category H227-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H373-H227-H302-H312-H315-H319-H335
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 206-718-2
RTECSNumber CZ5710000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only