missing translation for 'onlineSavingsMsg'
Learn More

1,2-Diethylbenzene, 97%

CAS: 135-01-3 | C10H14 | 134.222 g/mol

Supplier:  Thermo Scientific Chemicals L1877518

 View more versions of this product

Catalog No. AAL1877518


Only null left
Explore available promotions

Description

Description

1,2-Diethylbenzene is used as an intermediate and a building block in organic synthesis. 1,2-Diethylbenzene is used as one of the chemical composition of essential oils from flower and leaf of korean mint.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Diethylbenzene is used as an intermediate and a building block in organic synthesis. 1,2-Diethylbenzene is used as one of the chemical composition of essential oils from flower and leaf of korean mint.

Solubility
Miscible with alcohol, benzene and carbon tetrachloride. Immiscible with water.

Notes
Keep away from sources of ignition. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications

Specifications

1,2-Diethylbenzene
-31°C
183°C
C10H14
MFCD00009258
UN2049
o-diethylbenzene, benzene, 1,2-diethyl, benzene, diethyl, benzene, o-diethyl, diethylbenzol, unii-25hox6t1lu, 25hox6t1lu, diethylbenzenes mixed isomer, dsstox_cid_7866, dsstox_rid_78595
KVNYFPKFSJIPBJ-UHFFFAOYSA-N
1,2-diethylbenzene
8657
97%
135-01-3
0.88
55°C (131°F)
1.505
50 g
1904392
Miscible with alcohol,benzene and carbon tetrachloride. Immiscible with water.
CCC1=CC=CC=C1CC
134.222
134.22

Chemical Identifiers

135-01-3
134.222
KVNYFPKFSJIPBJ-UHFFFAOYSA-N
8657
CCC1=CC=CC=C1CC
C10H14
MFCD00009258
o-diethylbenzene, benzene, 1,2-diethyl, benzene, diethyl, benzene, o-diethyl, diethylbenzol, unii-25hox6t1lu, 25hox6t1lu, diethylbenzenes mixed isomer, dsstox_cid_7866, dsstox_rid_78595
1,2-diethylbenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c

H226-H304-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: DIETHYLBENZENE

EINECSNumber : 205-170-1

RTECSNumber : CZ5640000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only