missing translation for 'onlineSavingsMsg'
Learn More

1,2-Diethylbenzene, 97%

Catalog No. p-7047340
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1877504 2 g
AAL1877509 10 g
AAL1877518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1877504 Supplier Thermo Scientific Chemicals Supplier No. L1877504

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 135-01-3 | C10H14 | 134.222 g/mol

1,2-Diethylbenzene is used as an intermediate and a building block in organic synthesis. 1,2-Diethylbenzene is used as one of the chemical composition of essential oils from flower and leaf of korean mint.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Diethylbenzene is used as an intermediate and a building block in organic synthesis. 1,2-Diethylbenzene is used as one of the chemical composition of essential oils from flower and leaf of korean mint.

Solubility
Miscible with alcohol, benzene and carbon tetrachloride. Immiscible with water.

Notes
Keep away from sources of ignition. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 135-01-3
Molecular Formula C10H14
Molecular Weight (g/mol) 134.222
MDL Number MFCD00009258
InChI Key KVNYFPKFSJIPBJ-UHFFFAOYSA-N
Synonym o-diethylbenzene, benzene, 1,2-diethyl, benzene, diethyl, benzene, o-diethyl, diethylbenzol, unii-25hox6t1lu, 25hox6t1lu, diethylbenzenes mixed isomer, dsstox_cid_7866, dsstox_rid_78595
PubChem CID 8657
IUPAC Name 1,2-diethylbenzene
SMILES CCC1=CC=CC=C1CC

Specifications

Melting Point -31°C
Density 0.88
Boiling Point 183°C
Flash Point 55°C (131°F)
Refractive Index 1.505
Quantity 2 g
UN Number UN2049
Beilstein 1904392
Solubility Information Miscible with alcohol,benzene and carbon tetrachloride. Immiscible with water.
Formula Weight 134.22
Percent Purity 97%
Chemical Name or Material 1,2-Diethylbenzene
Show More Show Less
Hazard Category H226-H304-H315-H319-H335
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: DIETHYLBENZENE
EINECSNumber 205-170-1
RTECSNumber CZ5640000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only