Learn More
1,2-Diethylbenzene, 97%
CAS: 135-01-3 | C10H14 | 134.222 g/mol
Supplier: Thermo Scientific Chemicals L1877509
Description
1,2-Diethylbenzene is used as an intermediate and a building block in organic synthesis. 1,2-Diethylbenzene is used as one of the chemical composition of essential oils from flower and leaf of korean mint.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,2-Diethylbenzene is used as an intermediate and a building block in organic synthesis. 1,2-Diethylbenzene is used as one of the chemical composition of essential oils from flower and leaf of korean mint.
Solubility
Miscible with alcohol, benzene and carbon tetrachloride. Immiscible with water.
Notes
Keep away from sources of ignition. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications
| 1,2-Diethylbenzene | |
| -31°C | |
| 183°C | |
| C10H14 | |
| MFCD00009258 | |
| UN2049 | |
| o-diethylbenzene, benzene, 1,2-diethyl, benzene, diethyl, benzene, o-diethyl, diethylbenzol, unii-25hox6t1lu, 25hox6t1lu, diethylbenzenes mixed isomer, dsstox_cid_7866, dsstox_rid_78595 | |
| KVNYFPKFSJIPBJ-UHFFFAOYSA-N | |
| 1,2-diethylbenzene | |
| 8657 | |
| 97% |
| 135-01-3 | |
| 0.88 | |
| 55°C (131°F) | |
| 1.505 | |
| 10 g | |
| 1904392 | |
| Miscible with alcohol,benzene and carbon tetrachloride. Immiscible with water. | |
| CCC1=CC=CC=C1CC | |
| 134.222 | |
| 134.22 |
Chemical Identifiers
| 135-01-3 | |
| 134.222 | |
| KVNYFPKFSJIPBJ-UHFFFAOYSA-N | |
| 8657 | |
| CCC1=CC=CC=C1CC |
| C10H14 | |
| MFCD00009258 | |
| o-diethylbenzene, benzene, 1,2-diethyl, benzene, diethyl, benzene, o-diethyl, diethylbenzol, unii-25hox6t1lu, 25hox6t1lu, diethylbenzenes mixed isomer, dsstox_cid_7866, dsstox_rid_78595 | |
| 1,2-diethylbenzene |
Safety and Handling
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
H226-H304-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: DIETHYLBENZENE
EINECSNumber : 205-170-1
RTECSNumber : CZ5640000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only