missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichloropropane, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020621
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020621 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020621 Supplier Merck Emd Millipore Supplier No. 025771ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

1,2-Dichloropropane (1,2-D), is a colorless watery liquid with a sweet odor. It sinks in water and produces irritating vapor. It is a short-chain chlorinated aliphatic compound, commonly used as fumigant and as insecticide. It is a widespread groundwater contaminant. Anaerobic bacteria have the capacity to reductively dechlorinate it to propene. Featured Industries: Agriculture and Environmental

Specifications

CAS 78-87-5
Ignition Point 1035°F
Vapor Pressure 40 mmHg (19.4°C)
Vapor Density 3.89 (vs. air)
Density 1.156 g/mL (at 25°C)
Flash Point 15.0°C - closed cup
Linear Formula CH3 CHClCH2 Cl
MDL Number MFCD00000868
Melting Point -100°C
Boiling Point 95°C to 96°C
Synonym Propylene dichloride
Type Analytical Standard
Quantity 1 mL
Formula Weight 112.99
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less