missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichloro-4-fluorobenzene 98.0+%, TCI America™

Catalog No. D357025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D357025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D357025G Supplier TCI America Supplier No. D357025G
Only null left

Chemical Identifiers

CAS 1435-49-0
Molecular Formula C6H3Cl2F
Molecular Weight (g/mol) 164.99
MDL Number MFCD00018119
InChI Key QSDKXMVGRLVIQV-UHFFFAOYSA-N
Synonym 3,4-dichlorofluorobenzene, 3,4-dichloro-1-fluorobenzene, benzene, 1,2-dichloro-4-fluoro, pubchem3453, 3,4 dichlorofluorobenzene, acmc-1byo2, ksc494g9b, 1,2-dichloro-4-fluoro-benzene
PubChem CID 74028
IUPAC Name 1,2-dichloro-4-fluorobenzene
SMILES FC1=CC=C(Cl)C(Cl)=C1

Specifications

Melting Point -1°C
Color Yellow
Boiling Point 168°C
Quantity 25 g
Formula Weight 164.99
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Dichloro-4-fluorobenzene