missing translation for 'onlineSavingsMsg'
Learn More

(1,2-Dibromoethyl)benzene 98.0+%, TCI America™

Catalog No. D191425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D191425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D191425G Supplier TCI America Supplier No. D191425G

Chemical Identifiers

CAS 93-52-7
Molecular Formula C8H8Br2
Molecular Weight (g/mol) 263.96
MDL Number MFCD00000138
InChI Key SHKKTLSDGJRCTR-UHFFFAOYSA-N
Synonym 1,2-dibromoethyl benzene, dowspray 9, 1,2-dibromo-1-phenylethane, 1,2-dibromo phenyl ethane, benzene, 1,2-dibromoethyl, 1,2-dibromo-2-phenylethane, vic-styrene dibromide, alpha,beta-dibromoethylbenzene, ccris 2090, .alpha.,.beta.-dibromoethylbenzene
PubChem CID 7145
IUPAC Name 1,2-dibromoethylbenzene
SMILES C1=CC=C(C=C1)C(CBr)Br

Specifications

Melting Point 74°C
Color White-Yellow
Boiling Point 89°C
Quantity 25 g
UN Number 1759
Formula Weight 263.96
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (1,2-Dibromoethyl)benzene
RTECSNumber CZ1800100
TSCA Yes