missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromoethane, Restek

Catalog No. 06719267
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06719267 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06719267 Supplier Chromatographic Supplier No. RK 30272

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 106-93-4
Molecular Formula C2H4Br2
Molecular Weight (g/mol) 187.862
InChI Key PAAZPARNPHGIKF-UHFFFAOYSA-N
PubChem CID 7839
ChEBI CHEBI:28534
IUPAC Name 1,2-dibromoethane
SMILES C(CBr)Br

Specifications

Quantity 1 mL
Chemical Name or Material 1, 2-Dibromoethane