missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromoethane, 98%

Catalog No. p-7020717
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1276630 250 g
AAA127660B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAA1276630 Supplier Thermo Scientific Chemicals Supplier No. A1276630

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 106-93-4 | C2H4Br2 | 187.862 g/mol

1,2-dibromoethane is widely used as a bromine source in organic synthesis. It is utilized as a fumigant for treatment of logs for termites and beetles, for control of moths in beehives. It finds the application to make vinyl bromide, a precursor to some fire retardants. It is used as an intermediate in the preparation of dyes and pharmaceuticals. It is also used as a lead scavenger in antiknock mixtures added to gasolines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-dibromoethane is widely used as a bromine source in organic synthesis. It is utilized as a fumigant for treatment of logs for termites and beetles, for control of moths in beehives. It finds the application to make vinyl bromide, a precursor to some fire retardants. It is used as an intermediate in the preparation of dyes and pharmaceuticals. It is also used as a lead scavenger in antiknock mixtures added to gasolines.

Solubility
Miscible with alcohol, acetone, benzene, diethyl ether and ethyl acetate. Slightly miscible with water.

Notes
Special handling precautions required. View MSDS prior to purchase. MSDS are available online at www.alfa.com.

Chemical Identifiers

CAS 106-93-4
Molecular Formula C2H4Br2
Molecular Weight (g/mol) 187.862
MDL Number MFCD00000233
InChI Key PAAZPARNPHGIKF-UHFFFAOYSA-N
Synonym ethylene dibromide, ethylene bromide, sym-dibromoethane, ethane, 1,2-dibromo, alpha,beta-dibromoethane, bromuro di etile, 1,2-dibromaethan, 1,2-dibroomethaan, 1,2-ethylene dibromide, aadibroom
PubChem CID 7839
ChEBI CHEBI:28534
IUPAC Name 1,2-dibromoethane
SMILES C(CBr)Br

Specifications

Melting Point 9°C to 10°C
Density 2.179
Boiling Point 132°C
Flash Point None
Refractive Index 1.5385
Linear Formula BrCH2CH2Br
Quantity 250 g
UN Number UN1605
Sensitivity Light sensitive
Merck Index 14,3796
Solubility Information Miscible with alcohol,acetone,benzene,diethyl ether and ethyl acetate. Slightly miscible with water.
Formula Weight 187.87
Percent Purity 99%
Chemical Name or Material 1,2-Dibromoethane
Show More Show Less
Hazard Category H301+H311-H315-H319-H335-H350
Hazard Statement GHS H Statement
H301-H311-H331-H350-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: ETHYLENE DIBROMIDE
EINECSNumber 203-444-5
RTECSNumber KH9275000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only