missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromoethane, 99%

Catalog No. p-4258926
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC112790010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC112790010 Supplier Thermo Scientific Chemicals Supplier No. 112790010
Only null left

CAS: 106-93-4 | C2H4Br2 | 187.86 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 106-93-4
Molecular Formula C2H4Br2
Molecular Weight (g/mol) 187.86
InChI Key PAAZPARNPHGIKF-UHFFFAOYSA-N
Synonym ethylene dibromide, ethylene bromide, sym-dibromoethane, ethane, 1,2-dibromo, alpha,beta-dibromoethane, bromuro di etile, 1,2-dibromaethan, 1,2-dibroomethaan, 1,2-ethylene dibromide, aadibroom
PubChem CID 7839
ChEBI CHEBI:28534
IUPAC Name 1,2-dibromoethane
SMILES C(CBr)Br

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 9.0°C to 10.0°C
Color Colorless to Yellow
Density 2.1730g/mL
Boiling Point 131.0°C to 132.0°C
Flash Point >104°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5375 to 1.5395
Linear Formula BrCH2CH2Br
Quantity 1 L
Beilstein 01,9
Fieser 01,374; 04,126; 05,187; 12,570
Specific Gravity 2.173
Merck Index 15,385
Solubility Information Solubility in water: 4g/L (20°C). Other solubilities: miscible with most organic solvents
Formula Weight 187.86
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1,2-Dibromoethane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause cancer.
Toxic to aquatic life with long lasting effects.
Toxic in contact with skin.
Toxic if inhaled.
May cause respiratory irritation.
Toxic if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-444-5

RUO – Research Use Only