missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromobutane, 98%

CAS: 533-98-2 | C4H8Br2 | 215.92 g/mol

Supplier:  Thermo Scientific Chemicals B203560B

 View more versions of this product

Catalog No. AAB203560B


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1,2-Dibromobutane
-65°C
166°C to 167°C
C4H8Br2
MFCD00000157
1731380
butane, 1,2-dibromo, .alpha.-butylene dibromide, alpha-butylene dibromide, 1,2-dibomobutane, 1,2 dibromobutane, butane,2-dibromo, 1,2-dibromo-butane, alpha-butylene bromide, 1,2-butylene bromide, .alpha.-butylene bromide
CCC(Br)CBr
215.92
215.93
533-98-2
1.789
>110°C (230°F)
1.5141
1000 g
14,1566
CZWSZZHGSNZRMW-UHFFFAOYNA-N
1,2-dibromobutane
10792
98%

Chemical Identifiers

533-98-2
215.92
CZWSZZHGSNZRMW-UHFFFAOYNA-N
10792
CCC(Br)CBr
C4H8Br2
MFCD00000157
butane, 1,2-dibromo, .alpha.-butylene dibromide, alpha-butylene dibromide, 1,2-dibomobutane, 1,2 dibromobutane, butane,2-dibromo, 1,2-dibromo-butane, alpha-butylene bromide, 1,2-butylene bromide, .alpha.-butylene bromide
1,2-dibromobutane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 208-581-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only