missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromobenzene 97.0+%, TCI America™

Catalog No. p-7126068
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D016825G 25 g
D0168100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D016825G Supplier TCI America Supplier No. D016825G
Only null left

Chemical Identifiers

CAS 583-53-9
Molecular Formula C6H4Br2
Molecular Weight (g/mol) 235.906
MDL Number MFCD00000057
InChI Key WQONPSCCEXUXTQ-UHFFFAOYSA-N
Synonym o-dibromobenzene, dibromobenzene, benzene, 1,2-dibromo, ortho-dibromobenzene, benzene, o-dibromo, benzene, dibromo, unii-52k9w7u6eh, dibromobenzene mixed isomers, 1,2 dibromobenzene, 1,6-dibromobenzene
PubChem CID 11414
ChEBI CHEBI:37152
IUPAC Name 1,2-dibromobenzene
SMILES C1=CC=C(C(=C1)Br)Br

Specifications

Melting Point 6°C
Color Yellow
Boiling Point 224°C
Quantity 25 g
Formula Weight 235.91
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Dibromobenzene