missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromobenzene, 98%

Catalog No. p-3736123
Change view
Click to view available options
Quantity:
25 mL
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC348930250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC348930250 Supplier Thermo Scientific Chemicals Supplier No. 348930250
Only null left

CAS: 583-53-9 | C6H4Br2 | 235.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 583-53-9
Molecular Formula C6H4Br2
Molecular Weight (g/mol) 235.91
MDL Number MFCD00000057
InChI Key WQONPSCCEXUXTQ-UHFFFAOYSA-N
Synonym o-dibromobenzene, dibromobenzene, benzene, 1,2-dibromo, ortho-dibromobenzene, benzene, o-dibromo, benzene, dibromo, unii-52k9w7u6eh, dibromobenzene mixed isomers, 1,2 dibromobenzene, 1,6-dibromobenzene
PubChem CID 11414
ChEBI CHEBI:37152
IUPAC Name 1,2-dibromobenzene
SMILES C1=CC=C(C(=C1)Br)Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 4.0°C to 6.0°C
Color Colorless to Yellow or Pink
Density 1.9560g/mL
Boiling Point 224.0°C
Flash Point 91°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.6100 to 1.6120
Linear Formula C6H4Br2
Quantity 25 mL
Beilstein 05, 210
Specific Gravity 1.956
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with alcohol,acetone,ether,benzene,ca,rbon tetrachloride,n-heptane
Formula Weight 235.91
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1, 2-Dibromobenzene
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN
EINECSNumber 209-507-3

RUO – Research Use Only