missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromo-4-fluorobenzene, 98%

CAS: 2369-37-1 | C6H3Br2F | 253.896 g/mol

$59.66 - $199.48

Chemical Identifiers

CAS 2369-37-1
Molecular Formula C6H3Br2F
Molecular Weight (g/mol) 253.896
MDL Number MFCD00070780
InChI Key RNTGKISRXVFIIP-UHFFFAOYSA-N
PubChem CID 75402
IUPAC Name 1,2-dibromo-4-fluorobenzene
SMILES C1=CC(=C(C=C1F)Br)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2093588
View Documents
Thermo Scientific Chemicals
B2093588
2.5 g N/A
Only null left
 
AAB2093509
View Documents
Thermo Scientific Chemicals
B2093509
10 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2369-37-1
253.896
RNTGKISRXVFIIP-UHFFFAOYSA-N
1,2-dibromo-4-fluorobenzene
C6H3Br2F
MFCD00070780
75402
C1=CC(=C(C=C1F)Br)Br

Specifications

2369-37-1
MFCD00070780
3,4-dibromofluorobenzene, 3,4-dibromo-1-fluorobenzene, pubchem3445, acmc-1ccqu, intermediates-zcf02634, 1,2-dibromo-4-fluorobezene, benzene,1,2-dibromo-4-fluoro, 1,2-dibromo-4-fluorobenzene, benzene, 1,2-dibromo-4-fluoro
C1=CC(=C(C=C1F)Br)Br
253.896
253.91
1,2-Dibromo-4-fluorobenzene
C6H3Br2F
2.5 g
RNTGKISRXVFIIP-UHFFFAOYSA-N
1,2-dibromo-4-fluorobenzene
75402
98%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 219-131-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only