missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromo-4-fluorobenzene, 98%

Catalog No. AAB2093588
Change view
Click to view available options
Quantity:
2.5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2093588 2.5 g
AAB2093509 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2093588 Supplier Thermo Scientific Chemicals Supplier No. B2093588
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2369-37-1 | C6H3Br2F | 253.896 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2369-37-1
Molecular Formula C6H3Br2F
Molecular Weight (g/mol) 253.896
MDL Number MFCD00070780
InChI Key RNTGKISRXVFIIP-UHFFFAOYSA-N
PubChem CID 75402
IUPAC Name 1,2-dibromo-4-fluorobenzene
SMILES C1=CC(=C(C=C1F)Br)Br

Specifications

Quantity 2.5 g
Synonym 3,4-dibromofluorobenzene, 3,4-dibromo-1-fluorobenzene, pubchem3445, acmc-1ccqu, intermediates-zcf02634, 1,2-dibromo-4-fluorobezene, benzene,1,2-dibromo-4-fluoro, 1,2-dibromo-4-fluorobenzene, benzene, 1,2-dibromo-4-fluoro
Formula Weight 253.91
Percent Purity 98%
Chemical Name or Material 1,2-Dibromo-4-fluorobenzene
EINECSNumber 219-131-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only