missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Dibromo-4,5-dimethoxybenzene 98.0+%, TCI America™
Supplier: TCI America D501925G
Specifications
| 1,2-Dibromo-4,5-dimethoxybenzene | |
| 89°C | |
| C8H8Br2O2 | |
| 25 g | |
| ZYCLQXMMFJREPJ-UHFFFAOYSA-N | |
| 1,2-dibromo-4,5-dimethoxybenzene | |
| 2758033 | |
| ≥98.0% (GC) |
| 37895-73-1 | |
| White-Yellow | |
| MFCD00014894 | |
| 4,5-dibromoveratrole, 4,5-dibromoveratrol, benzene, 1,2-dibromo-4,5-dimethoxy, acmc-1aeu5, amth147, 1.2-dibromo-4.5-dimethoxy-benzene, 1,2-bis bromanyl-4,5-dimethoxy-benzene | |
| COC1=CC(=C(C=C1OC)Br)Br | |
| 295.958 | |
| 295.96 | |
| Crystalline Powder |
Chemical Identifiers
| 37895-73-1 | |
| 295.958 | |
| ZYCLQXMMFJREPJ-UHFFFAOYSA-N | |
| 2758033 | |
| COC1=CC(=C(C=C1OC)Br)Br |
| C8H8Br2O2 | |
| MFCD00014894 | |
| 4,5-dibromoveratrole, 4,5-dibromoveratrol, benzene, 1,2-dibromo-4,5-dimethoxy, acmc-1aeu5, amth147, 1.2-dibromo-4.5-dimethoxy-benzene, 1,2-bis bromanyl-4,5-dimethoxy-benzene | |
| 1,2-dibromo-4,5-dimethoxybenzene |
Safety and Handling
TSCA : No