missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromo-4,5-dimethoxybenzene 98.0+%, TCI America™

Catalog No. D50195G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50195G 5 g
D501925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D50195G Supplier TCI America Supplier No. D50195G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 37895-73-1
Molecular Formula C8H8Br2O2
Molecular Weight (g/mol) 295.958
MDL Number MFCD00014894
InChI Key ZYCLQXMMFJREPJ-UHFFFAOYSA-N
Synonym 4,5-dibromoveratrole, 4,5-dibromoveratrol, benzene, 1,2-dibromo-4,5-dimethoxy, acmc-1aeu5, amth147, 1.2-dibromo-4.5-dimethoxy-benzene, 1,2-bis bromanyl-4,5-dimethoxy-benzene
PubChem CID 2758033
IUPAC Name 1,2-dibromo-4,5-dimethoxybenzene
SMILES COC1=CC(=C(C=C1OC)Br)Br

Specifications

Melting Point 89°C
Color White-Yellow
Quantity 5 g
Formula Weight 295.96
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Dibromo-4,5-dimethoxybenzene