missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromo-3,4,5,6-tetramethylbenzene, 98%

Catalog No. p-7032186
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3355803 1 g
AAH3355806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3355803 Supplier Thermo Scientific Chemicals Supplier No. H3355803

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 36321-73-0 | C10H12Br2 | 292.014 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 36321-73-0
Molecular Formula C10H12Br2
Molecular Weight (g/mol) 292.014
MDL Number MFCD00154987
InChI Key VEZJOZSIGSNYEB-UHFFFAOYSA-N
Synonym benzene,1,2-dibromo-3,4,5,6-tetramethyl, pubchem21478, benzene, 1,2-dibromo-3,4,5,6-tetramethyl, 1,2-bis bromanyl-3,4,5,6-tetramethyl-benzene
PubChem CID 2752606
IUPAC Name 1,2-dibromo-3,4,5,6-tetramethylbenzene
SMILES CC1=C(C(=C(C(=C1C)Br)Br)C)C

Specifications

Quantity 1 g
Formula Weight 292.01
Percent Purity 98%
Chemical Name or Material 1,2-Dibromo-3,4,5,6-tetramethylbenzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only