missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibenzoylethylene, predominantly trans, 96%

Catalog No. AAL1264306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1264306 5 g
AAL1264303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1264306 Supplier Thermo Scientific Chemicals Supplier No. L1264306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 959-28-4 | C16H12O2 | 236.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 959-28-4
Molecular Formula C16H12O2
Molecular Weight (g/mol) 236.27
MDL Number MFCD00003083
InChI Key WYCXGQSQHAXLPK-VAWYXSNFSA-N
Synonym trans-1,2-dibenzoylethylene, dibenzoylethylene, 1,2-dibenzoylethylene, 1,4-diphenyl-2-butene-1,4-dione, trans-1,4-diphenyl-2-butene-1,4-dione, 1,2-dibenzoylethene, ethylene, 1,2-dibenzoyl, usaf nd-57, 2-butene-1,4-dione, 1,4-diphenyl, trans-diphenacylidene
PubChem CID 735960
IUPAC Name (E)-1,4-diphenylbut-2-ene-1,4-dione
SMILES C1=CC=C(C=C1)C(=O)C=CC(=O)C2=CC=CC=C2

Specifications

Melting Point 103°C to 108°C
Quantity 5 g
Beilstein 743958
Formula Weight 236.27
Percent Purity 96%
Chemical Name or Material 1,2-Dibenzoylethylene, predominantly trans
EINECSNumber 213-498-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only