Learn More
1,2-Diaminocyclohexane, 99%, mixture of cis and trans
CAS: 694-83-7 | C6H14N2 | 114.19 g/mol
Supplier: Thermo Scientific Chemicals 158100010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
| 1, 2-Diaminocyclohexane | |
| 694-83-7 | |
| 100.0 | |
| 0.9300g/mL | |
| 75°C | |
| 98.5% min. (GC) | |
| C6H14N2 | |
| C6H10(NH2)2 | |
| 1 kg | |
| 0.93 | |
| Solubility in water: miscible. | |
| C1CCC(C(C1)N)N | |
| 114.19 | |
| 7.2 mPa.s (21°C) | |
| 99% | |
| Liquid |
| tech., 90% | |
| 98.5 | |
| 2.0°C to 15.0°C | |
| 188.0°C to 192.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4860 to 1.4920 | |
| MFCD00001491 | |
| 13, 1 | |
| 1,2-cyclohexanediamine, 1,2-diaminocyclohexane, 1,2-cylohexanediamine, cyclohex-1,2-ylenediamine, 1r,2r---1,2-diamino cyclohexane, dsstox_cid_7301, dsstox_rid_78396, dsstox_gsid_27301, +/--trans-1,2-diaminocyclohexane, cis-1,2-cyclohexandiamine | |
| SSJXIUAHEKJCMH-UHFFFAOYSA-N | |
| cyclohexane-1,2-diamine | |
| 4610 | |
| 114.19 | |
| Technical |
Identifiants chimiques
| 694-83-7 | |
| 114.19 | |
| SSJXIUAHEKJCMH-UHFFFAOYSA-N | |
| 4610 | |
| C1CCC(C(C1)N)N |
| C6H14N2 | |
| MFCD00001491 | |
| 1,2-cyclohexanediamine, 1,2-diaminocyclohexane, 1,2-cylohexanediamine, cyclohex-1,2-ylenediamine, 1r,2r---1,2-diamino cyclohexane, dsstox_cid_7301, dsstox_rid_78396, dsstox_gsid_27301, +/--trans-1,2-diaminocyclohexane, cis-1,2-cyclohexandiamine | |
| cyclohexane-1,2-diamine |
Sécurité et manipulation
GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 211-776-7
missing translation for 'rtecsNumber' : GU8749500
missing translation for 'tsca' : TSCA
RUO – Research Use Only