missing translation for 'onlineSavingsMsg'
Learn More

1,2-Diaminocyclohexane, 99%, mixture of cis and trans

Catalog No. p-100009
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC158102500 250 g Glass bottle
AC158100010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC158102500 Supplier Thermo Scientific Chemicals Supplier No. 158102500

CAS: 694-83-7 | C6H14N2 | 114.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 694-83-7
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.19
MDL Number MFCD00001491
InChI Key SSJXIUAHEKJCMH-UHFFFAOYSA-N
Synonym 1,2-cyclohexanediamine, 1,2-diaminocyclohexane, 1,2-cylohexanediamine, cyclohex-1,2-ylenediamine, 1r,2r---1,2-diamino cyclohexane, dsstox_cid_7301, dsstox_rid_78396, dsstox_gsid_27301, +/--trans-1,2-diaminocyclohexane, cis-1,2-cyclohexandiamine
PubChem CID 4610
IUPAC Name cyclohexane-1,2-diamine
SMILES C1CCC(C(C1)N)N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 2.0°C to 15.0°C
Density 0.9300g/mL
Boiling Point 188.0°C to 192.0°C
Flash Point 75°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4860 to 1.4920
Linear Formula C6H10(NH2)2
Quantity 250 g
Beilstein 13, 1
Specific Gravity 0.93
Solubility Information Solubility in water: miscible.
Viscosity 7.2 mPa.s (21°C)
Formula Weight 114.19
Percent Purity 99%
Grade Technical
Physical Form Liquid
Chemical Name or Material 1, 2-Diaminocyclohexane, tech., 90%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 211-776-7
RTECSNumber GU8749500
TSCA TSCA

RUO – Research Use Only