missing translation for 'onlineSavingsMsg'
Learn More

1,2-Diamino-4,5-difluorobenzene, 97%

Catalog No. p-4812558
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC449810010 1 g
AC449810050 5 g
AC449810250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC449810010 Supplier Thermo Scientific Chemicals Supplier No. 449810010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 76179-40-3 | C6H6F2N2 | 144.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 76179-40-3
Molecular Formula C6H6F2N2
Molecular Weight (g/mol) 144.13
MDL Number MFCD00061131
InChI Key PPWRHKISAQTCCG-UHFFFAOYSA-N
Synonym 1,2-diamino-4,5-difluorobenzene, 4,5-difluoro-1,2-phenylenediamine, 4,5-difluoro-o-phenylenediamine, 2-amino-4,5-difluorophenylamine, 4,5-difluoro-benzene-1,2-diamine, 4,5-difluoro-1,2-benzenediamine, 1,2-benzenediamine,4,5-difluoro, 1,2-benzenediamine, 4,5-difluoro, pubchem4354, 2-amino-4,5-difluoroaniline
PubChem CID 2736755
IUPAC Name 4,5-difluorobenzene-1,2-diamine
SMILES NC1=CC(F)=C(F)C=C1N

Specifications

Melting Point 130°C to 133°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (F)C6H2(NH2)2
Quantity 1 g
Formula Weight 144.12
Percent Purity 97%
Chemical Name or Material 1, 2-Diamino-4, 5-difluorobenzene
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only