missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Diamino-2-methylpropane 97.0+%, TCI America™
Supplier: TCI America D210225ML
Specifications
| 1,2-Diamino-2-methylpropane | |
| Yellow | |
| C4H12N2 | |
| 25 mL | |
| 2-Methyl-1,2-propanediamine | |
| CC(C)(CN)N | |
| 88.154 | |
| 88.15 | |
| Liquid |
| 811-93-8 | |
| 52°C | |
| MFCD00008054 | |
| 2734 | |
| OPCJOXGBLDJWRM-UHFFFAOYSA-N | |
| 2-methylpropane-1,2-diamine | |
| 13128 | |
| ≥97.0% (GC,T) |
Chemical Identifiers
| 811-93-8 | |
| 88.154 | |
| OPCJOXGBLDJWRM-UHFFFAOYSA-N | |
| 13128 | |
| CC(C)(CN)N |
| C4H12N2 | |
| MFCD00008054 | |
| 2-Methyl-1,2-propanediamine | |
| 2-methylpropane-1,2-diamine |
Safety and Handling
RTECSNumber : TX8250000
TSCA : No